Learn More
Emma Safety Footwear Zolder Safety Shoes
Water-repellant oiled Nubuck uppers with internal steel toe protection and an exceptional 'sticky' grip, even on wet and greasy surfaces.
Brand: Emma Safety Footwear MM50884646
This item is not returnable.
View return policy
Description
- S3-Safety shoe with sole plate for wet work
- GripForce™ Duo/HRO
- Oiled Nubuck
- Heat Resistant Outsole
Specifications
EN ISO 20345:2011 | |
Zolder | |
Oiled Nubuck |
Brown | |
Full grain Nappa leather / Split leather | |
46 |
Product Content Correction
Product Title
Emma Safety Footwear Zolder Safety Shoes > Size: 46