Learn More
Emma Safety Footwear Zolder Safety Shoes
Water-repellant oiled Nubuck uppers with internal steel toe protection and an exceptional 'sticky' grip, even on wet and greasy surfaces.
Brand:  Emma Safety Footwear MM50884643
 View more versions of this product
Product Code. 15908651
  • 1660.00 NOK / 1 pair
Estimated Shipment: 10-07-2025
to see stock.
Explore more special offers
Description
  • S3-Safety shoe with sole plate for wet work
  • GripForce™ Duo/HRO
  • Oiled Nubuck
  • Heat Resistant Outsole
Specifications
Specifications
EN ISO 20345:2011
Zolder
Oiled Nubuck
Brown
Full grain Nappa leather / Split leather
43
Product Title
Emma Safety Footwear Zolder Safety Shoes > Size: 43