Learn More
ZNF131 293T Cell Overexpression Lysate (Denatured), Abnova
Brand:  Abnova H00007690-T01.100uL
Product Code. 16112042
  • 2635.00 NOK / 100µL
Explore more special offers
Description
Description
Western Blotting
Specifications
Specifications
293T
ZNF131 293T Cell Transient Overexpression Lysate(Denatured)
56.21
pCMV-ZNF131 full-length
100 μL
Human
Western Blot
BC035875.2
6.8
Transient overexpression cell Lysate was tested with Anti-ZNF131 antibody by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.