Learn More
Zirconium Standard Metal Solution 1000ppm, for AAS, Solution HCl 1M, Fisher Chemical
Click to view available options
Quantity:
500 mL
Packaging:
HDPE plastic bottle
Specifications
Specifications
Packaging | HDPE plastic bottle |
Quantity | 500 mL |
Color | Colorless |
MDL Number | 11303 |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Zirconium solution 1000 ppm