Learn More
Zirconium oxide Yttrium oxide substrate, 10x10x1mm, polished one side, 100 orientation, Thermo Scientific Chemicals
Product Code. 11308867
Click to view available options
Quantity:
1 Ea.
5 Ea.
Unit Size:
Each
Pack of 5
Product Code. 11308867
Brand: Thermo Scientific Alfa Aesar 038509.KT
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Zirconium Oxide Yttrium Oxide Substrate
Name Note Polished One Side
Physical Form Plate
Quantity 1 Ea.
Molecular Formula ZrO2 + 9.5% Y2O3
MDL Number MFCD01868885