missing translation for 'onlineSavingsMsg'
Learn More
Zinc selenide, 99.995% (metals basis) Product Code.: 11336688

Zinc selenide, 99.995% (metals basis)

Product Code. p-7049213
50 g, 50g
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11336688

Brand: Thermo Scientific Alfa Aesar 013241.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1315-09-9 | SeZn | 144.35 g/mol

Zinc selenide is used to form II-VI light-emitting diodes and diode lasers. It emits blue light. It is doped with chromium (ZnSe:Cr) has been used as an infrared laser gain medium. It is used as scintillator with emission peak at 640 nm. It is used in x-ray and gamma ray detectors. It is used as an infrared optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315-09-9
Molecular Formula SeZn
Molecular Weight (g/mol) 144.35
MDL Number MFCD00011296
InChI Key GTLQJUQHDTWYJC-UHFFFAOYSA-N
Synonym zinc selenide znse, znse, zinc selenide, spectro, zinc selenide, powder, 10 mum trace metals basis, zinc selenide crystal optic disc, 13mm x 1mm, polished both sides, zinc selenide crystal optic disc, 13mm x 2mm, polished both sides, zinc selenide crystal optic disc, 25mm x 2mm, polished both sides, zinc selenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick, zinc selenide optical window, 12.7mm 0.5in dia x 2mm 0.08in thick, zinc selenide optical window, 25.4mm 1.0in dia x 1mm 0.04in thick
IUPAC Name zinc(2+) selandiide
SMILES [Zn++].[Se--]

Specifications

Melting Point >1100°C
Physical Form Lump
Quantity 50 g
UN Number UN3283
Merck Index 14,10156
Solubility Information Insoluble in water. Decomposes in dilute nitric acid.
Formula Weight 144.33
Percent Purity 99.995%
Odor Odorless
Sensitivity Air Sensitive
Density 5.42 g/mL at 15°C
Assay (metals basis)
Chemical Name or Material Zinc selenide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number442143-000 Certificate TypeCertificate of Analysis Date14/06/2025 Product Code
Lot Number433955-000 Certificate TypeCertificate of Analysis Date14/06/2025 Product Code
Lot Number442143-000 Certificate TypeCertificate of Analysis Date24/04/2025 Product Code
Lot Number433955-000 Certificate TypeCertificate of Analysis Date24/04/2025 Product Code
Lot NumberT16B035 Certificate TypeCertificate of Analysis Date01/02/2025 Product Code013241.18,013241.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zinc selenide, 99.995% (metals basis) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.