missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Perchlorate Hexahydrate, MP Biomedicals
Click to view available options
Quantity:
100 g
Specifications
Specifications
CAS | 10025-64-6 |
Quantity | 100 g |
Molecular Formula | Cl2H12O14Zn |
Synonym | zinc perchlorate hexahydrate, perchloric acid, zinc salt, hexahydrate, 2clo4.zn.6h2o, zinc diperchlorate hexahydrate, zinc perchlorate 6h2o, zincperchloratehexahydrate, zinc 2+ hexahydrate diperchlorate, zinc perchlorate hexahydrate, reagent grade, zinc, ion zn2+ hexahydrate diperchlorate ion, zinc perchlorate hexahydrate trace metals basis 10g |
InChI Key | PADPILQDYPIHQQ-UHFFFAOYSA-L |
SMILES | O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Zn+2] |
IUPAC Name | zinc;diperchlorate;hexahydrate |
Molecular Weight (g/mol) | 372.362 |
PubChem CID | 202253 |
Formula Weight | 372.4 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'