Learn More
Thermo Scientific Chemicals YP Base Medium
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Description
For maintaining and developing yeast in molecular microbiology procedures
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Form | Powder |
Packaging Type | Plastic Bottle |
Quantity | 100 g |
Sensitivity | Hygroscopic; Moisture sensitive |
Product Title
Thermo Scientific Chemicals YP Base Medium >