missing translation for 'onlineSavingsMsg'
Learn More

YM 022, Tocris Bioscience™

Product Code. 30001246
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly potent, selective non-peptide CCK2 antagonist

Extremely potent and highly selective non-peptide CCK2 silent antagonist (Ki values are 68 pM and 63 nM at CCK2 and CCK1 receptors respectively). Acts in vivo to potently inhibit gastrin-induced gastric acid secretion and histidine decarboxylase activation with a long duration of action.

Chemical Identifiers

CAS 145084-28-2
Molecular Formula C32H28N4O3
Molecular Weight (g/mol) 516.601
InChI Key YCXFHPUBGMMWJQ-PMERELPUSA-N
Synonym unii-772cp7w12n, 1-2,3-dihydro-1-2'-methylphenacyl-2-oxo-5-phenyl-1h-1,4-benzodiazepin-3-yl-3-3-methylphenyl urea, 3-3-methylphenyl-1-3r-1-2-2-methylphenyl-2-oxoethyl-2-oxo-5-phenyl-3h-1,4-benzodiazepin-3-yl urea, urea, n-2,3-dihydro-1-2-2-methylphenyl-2-oxoethyl-2-oxo-5-phenyl-1h-1,4-benzodiazepin-3-yl-n'-3-methylphenyl-, r, urea,n-3r-2,3-dihydro-1-2-2-methylphenyl-2-oxoethyl-2-oxo-5-phenyl-1h-1,4-benzodiazepin-3-yl-n'-3-methylphenyl, r-n-2,3-dihydro-1-2-2-methylphenyl-2-oxoethyl-2-oxo-5-phenyl-1h-1,4-benzodiazepin-3-yl-n'-3-methylphenyl-urea, ym022 racemate, d05hyl, gtpl888, ym hplc
PubChem CID 122130
IUPAC Name 1-(3-methylphenyl)-3-[(3R)-1-[2-(2-methylphenyl)-2-oxoethyl]-2-oxo-5-phenyl-3H-1,4-benzodiazepin-3-yl]urea
SMILES CC1=CC(=CC=C1)NC(=O)NC2C(=O)N(C3=CC=CC=C3C(=N2)C4=CC=CC=C4)CC(=O)C5=CC=CC=C5C

Specifications

Quantity 10 mg
Formula Weight 516.6
Percent Purity >99%
Chemical Name or Material YM 022
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.