missing translation for 'onlineSavingsMsg'
Learn More

YM 022, Tocris Bioscience™

Product Code. 18043822
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly potent, selective non-peptide CCK2 antagonist

Extremely potent and highly selective non-peptide CCK2 silent antagonist (Ki values are 68 pM and 63 nM at CCK2 and CCK1 receptors respectively). Acts in vivo to potently inhibit gastrin-induced gastric acid secretion and histidine decarboxylase activation with a long duration of action.

Chemical Identifiers

CAS 145084-28-2
Molecular Formula C32H28N4O3
Molecular Weight (g/mol) 516.601
InChI Key YCXFHPUBGMMWJQ-PMERELPUSA-N
Synonym unii-772cp7w12n, 1-2,3-dihydro-1-2'-methylphenacyl-2-oxo-5-phenyl-1h-1,4-benzodiazepin-3-yl-3-3-methylphenyl urea, 3-3-methylphenyl-1-3r-1-2-2-methylphenyl-2-oxoethyl-2-oxo-5-phenyl-3h-1,4-benzodiazepin-3-yl urea, urea, n-2,3-dihydro-1-2-2-methylphenyl-2-oxoethyl-2-oxo-5-phenyl-1h-1,4-benzodiazepin-3-yl-n'-3-methylphenyl-, r, urea,n-3r-2,3-dihydro-1-2-2-methylphenyl-2-oxoethyl-2-oxo-5-phenyl-1h-1,4-benzodiazepin-3-yl-n'-3-methylphenyl, r-n-2,3-dihydro-1-2-2-methylphenyl-2-oxoethyl-2-oxo-5-phenyl-1h-1,4-benzodiazepin-3-yl-n'-3-methylphenyl-urea, ym022 racemate, d05hyl, gtpl888, ym hplc
PubChem CID 122130
IUPAC Name 1-(3-methylphenyl)-3-[(3R)-1-[2-(2-methylphenyl)-2-oxoethyl]-2-oxo-5-phenyl-3H-1,4-benzodiazepin-3-yl]urea
SMILES CC1=CC(=CC=C1)NC(=O)NC2C(=O)N(C3=CC=CC=C3C(=N2)C4=CC=CC=C4)CC(=O)C5=CC=CC=C5C

Specifications

Quantity 50 mg
Formula Weight 516.6
Percent Purity >99%
Chemical Name or Material YM 022
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'