missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YES1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20243-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to YES1.
YES is a member of the Src family of non-receptor tyrosine kinases participating in a wide range of cellular signalling pathways. The gene that encodes YES is the cellular homolog of the Yamaguchi sarcoma virus oncogene. This gene lies in proximity to thymidylate synthase gene on chromosome 18, and a pseudogene of it has mapped to chromosome 22.Specifications
YES1 | |
Polyclonal | |
Unconjugated | |
YES1 | |
c yes, HsT441, P61 YES, Proto oncogene c Yes, Yes, YES1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22612, 24884, 7525 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
F1LM93, P07947, Q04736 | |
YES1 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction