Learn More
Yeast-Torula
For biological research purposes.
Brand: Thermo Scientific J23551.Q1
This item is not returnable.
View return policy
Description
- Format: Solid
Specifications
Yeast-Torula | |
Solid | |
25 lb. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
234631 | Certificate of Analysis | 17/10/2023 | J23551.Q1,J23551.Q5 |
233121 | Certificate of Analysis | 17/10/2023 | J23551.Q1,J23551.Q5 |
230256 | Certificate of Analysis | 22/03/2023 | J23551.Q1,J23551.Q5 |
180681 | Certificate of Analysis | 27/11/2022 | J23551.Q1,J23551.Q5 |
179236 | Certificate of Analysis | 27/11/2022 | J23551.Q1,J23551.Q5 |
Product Content Correction
Product Title
Yeast-Torula > 25 lb.