Learn More
Thermo Scientific™ Xpect™ Rotavirus Test
Rapidly detect Rotavirus from human fecal samples.
Brand: Thermo Scientific™ R24655
5428.30 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Optimize your workflow by rapidly detecting rotavirus antigen in human fecal specimens using Thermo Scientific™ Xpect Rotavirus Test. Combine excellent clinical performance with a simple, two-step procedure that offers accurate results within 15 minutes.
Use this rapid, lateral flow immunoassay for the direct qualitative detection of Rotavirus antigen. Enhance laboratory performance and efficiency with excellent clinical sensitivity and specificity and a simple walk-away procedure.- Clearly visible, easy-to-read results in just 15 minutes
- Less than one minute hands-on time
- 100% sensitivity and specificity
- Stock up with 18-month shelf life at room-temperature storage
Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.
Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Rotavirus Immunoassay | |
20 Tests/Kit |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Xpect Rotavirus
File size: 425.1 kB
Product Content Correction
Product Title
Thermo Scientific™ Xpect™ Rotavirus Test > Rotavirus Immunoassay; 20 tests/kit