Learn More
Xanthine, 98%, Thermo Scientific Chemicals
Product Code. 10309233
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69-89-6
Molecular Formula C5H4N4O2
Molecular Weight (g/mol) 152.11
MDL Number MFCD00078453
InChI Key LRFVTYWOQMYALW-UHFFFAOYSA-N
Synonym xanthine, 2,6-dihydroxypurine, xanthin, 2,6-dioxopurine, pseudoxanthine, 1h-purine-2,6 3h,7h-dione, isoxanthine, xanthic oxide, 1h-purine-2,6-diol, 9h-purine-2,6-diol
PubChem CID 1188
ChEBI CHEBI:17712
IUPAC Name 3,7-dihydropurine-2,6-dione
SMILES C1=NC2=C(N1)C(=O)NC(=O)N2
Specifications
Melting Point >300.0°C
Color White to Yellow
Loss on Drying 1% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 26, 447
Merck Index 15, 10257
Solubility Information Solubility in water: 1g/14.5l (16°C). Other solubilities: soluble in potassium hydroxide and mineral acids,practically insoluble in most common organic,solvents
Spectrophotometry 98 to 102% (lambda max. 278nm,pH 10) molar absorptivity 9300+/-2%,on dry basis
Formula Weight 152.11
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Xanthine
Show More Show Less
compliance-icons
Product Identifier
  • Xanthine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.