Learn More
Fisher Chemical X25 Sodium sulphate, anhydrous, PLV. SLR
Brand:  Fisher Chemical S/6600/70
4644.42 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
X25 Sodium sulphate, anhydrous, PLV. SLR
Product Code. 10831471
  • 4715.00 NOK / 25kg
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2537835 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberS/6600/70,S/6600/60,S/6600/71,S/6600/63,S/6600/53,S/6600/65
Lot Number2530306 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberS/6600/70,S/6600/60,S/6600/71,S/6600/63,S/6600/53,S/6600/65
Lot Number2527238 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberS/6600/70,S/6600/60,S/6600/71,S/6600/63,S/6600/53,S/6600/65
Lot Number2527237 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberS/6600/70,S/6600/60,S/6600/71,S/6600/63,S/6600/53,S/6600/65
Lot Number2527238 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog NumberS/6600/70,S/6600/71
Promotions
Promotions
Product Content Correction
Product Title
Fisher Chemical X25 Sodium sulphate, anhydrous, PLV. SLR >