Learn More
Fisherbrand X100 1.5ml Short Thread Vial, 32 x 11.6mm, amber
Brand:  Fisherbrand 11 09 3149
81.09 NOK valid until 2025-05-31
Use promo code "24127" to get your promotional price.
X100 1.5ml Short Thread Vial, 32 x 11.6mm, amber
Product Code. 15946001
  • 91.05 NOK / Pack of 100
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Documents
One moment while we fetch your results.
Product Title
Fisherbrand X100 1.5ml Short Thread Vial, 32 x 11.6mm, amber >