Learn More
Kartell™ Polypropylene Watch Glasses
Use a watch glass to evaporate liquids, hold solids when weighing a sample, and in situations where cross-contamination could occur. Kartell™ Polypropylene Watch Glass has good heat and chemical resistance.
Brand: Kartell™ 0045400
196.82 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- Very stable
- Moulded base
- Can withstand temperatures up to 120°C
Product Content Correction
Product Title
Kartell™ Polypropylene Watch Glasses > DiameterMetric: 80mm