Learn More
Kartell™ Polypropylene Watch Glasses
Use a watch glass to evaporate liquids, hold solids when weighing a sample, and in situations where cross-contamination could occur. Kartell™ Polypropylene Watch Glass has good heat and chemical resistance.
Brand:  Kartell™ 0045600
Product Code. 10448842
  • 351.00 NOK / Pack of 10
Only null left
Estimated Shipment: 30-06-2025
Description
  • Very stable
  • Moulded base
  • Can withstand temperatures up to 120°C
Specifications
Specifications
Weight: 15g
Round
1.25 mm
118.5 mm
Polypropylene
Watch Glass
Up to 120°C
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kartell™ Polypropylene Watch Glasses > DiameterMetric: 125mm