Learn More
Lenz Laborglasintrumente X10 Test tube w/ NS joint, w/o graduation, w/ PE stopper, socket NS 24/29, Ø 28xheight 150mm,
Brand: Lenz Laborglasintrumente 05700138
X10 Test tube w/ NS joint, w/o graduation, w/ PE stopper, socket NS 24/29, Ø 28xheight 150mm,
This item is not returnable.
View return policy
Product Content Correction
Product Title
Lenz Laborglasintrumente X10 Test tube w/ NS joint, w/o graduation, w/ PE stopper, socket NS 24/29, Ø 28xheight 150mm, >