Learn More
Heathrow Scientific x-tourn™ Tourniquets
x-tourn™ Tourniquets are high-quality, eliminating roping or curling when used. Latex free, no rubber proteins, resins or fatty acids that can produce hypersensitive reactions.
Brand: Heathrow Scientific HS120111
179.45 NOK valid until 2024-12-31
Use promo code "21693" to get your promotional price.
This item is not returnable.
View return policy
Description
- Less abrasive than other brands, uniform thickness prevents shredding and breaking
- Made from supple amd smooth polyisoprene
- Latex free
- Disposable
Specifications
Orange | |
Polyisoprene | |
Single-use | |
18 in. | |
2.54 cm |
Yes | |
25/Roll | |
45.7 cm | |
1 in. |