Learn More
WTW™ WTW™ Rectangular Cuvette
For automatic detection of the photLab™ filter and spectral photometer
Brand: WTW™ 250614
1497.44 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
The glass and quartz cuvettes with matte side surface are suitable for the automatic cuvette detection of the photoLab™ filter and spectral photometer. When using disposable cuvettes, the material PMMA as well as a structured or matte side surface is required due to the cuvette detection by the photoLab™ photometers. The glass and quartz cuvettes have a marking in order to place the cuvettes into the photometer in the same direction for an even higher precision.Cuvettes for turbidity measurement can be marked to prevent influences by glass: This allows a higher accuracy, also for values < 1 NTU.
Specifications
Cuvette | |
50 mm | |
Rectangle |
Product Content Correction
Product Title
WTW™ WTW™ Rectangular Cuvette > Number of Curvette: 1; Pathlength: 50mm