Learn More
WTW™ WTW™ Rectangular Cuvette
For automatic detection of the photLab™ filter and spectral photometer
Brand:  WTW™ 250600
1754.82 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10324781
  • 1835.00 NOK / Each
Only null left
In Stock
Add to basket
Description
Description
The glass and quartz cuvettes with matte side surface are suitable for the automatic cuvette detection of the photoLab™ filter and spectral photometer. When using disposable cuvettes, the material PMMA as well as a structured or matte side surface is required due to the cuvette detection by the photoLab™ photometers. The glass and quartz cuvettes have a marking in order to place the cuvettes into the photometer in the same direction for an even higher precision.Cuvettes for turbidity measurement can be marked to prevent influences by glass: This allows a higher accuracy, also for values < 1 NTU.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cuvette
20 mm
Rectangle
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
WTW™ WTW™ Rectangular Cuvette > Number of Curvette: 2; Pathlength: 20mm