missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wortmannin, >99% purity, For TLC, HPLC analysis, 97.96%, MP Biomedicals™
Click to view available options
Quantity:
1 mg
5 mg
Specifications
Specifications
Color | White |
Assay Percent Range | 0.9796 |
Quantity | 5 mg |
Formula Weight | 428.4 |
Percent Purity | >99% |
Physical Form | Powder |
Chemical Name or Material | Wortmannin, For TLC and HPLC analysis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction