Learn More
WTW™ Wireless Module For Plug Head Sensor
The Wireless Module For Plug Head Sensor is for use with the WTW™ range of IDS Intelligent Digital Sensors
Brand: WTW™ 108141
2105.78 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Wireless modules for IDS plug head sensors are simply plugged onto the sensor and automatically connect to the meter, same module for all parameters
- Safe and simple with no cables
- Freedom to operate where a meter would usually be in the way
- Even with portable field measurements under difficult conditions, one hand is always free
- Recording measurement values by simply pressing a button on the sensor module
Specifications
Re-chargeable LiPo battery 230 mAh | |
Splash water protected to IP 66 | |
WTS™ IDS Plug Head Sensors for Multi-Parameter Meters |