Learn More
R&L Enterprises™ Wire Gauze with Ceramic Center
Provides excellent support to beakers and other glassware during heating on ring stands. R&L Enterprises Wire Gauze with Ceramic Center is a flat square manufactured from galvanized iron mesh.
Brand:  R&L Enterprises™ 9073
Product Code. 12988950
  • 333.00 NOK / Pack of 10
Estimated Shipment: 03-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Non-fraying edges
  • Durable and long-lasting
  • Ceramic center disperses heat evenly
Specifications
Specifications
Ceramic Center
Non-Fraying
10 Pack
150 mm
Wire Gauze
150 x 150 mm
Ceramic center
Square
Iron
150 mm
Product Title
R&L Enterprises™ Wire Gauze with Ceramic Center > Dimensions: 150 x 150mm