Learn More
Buerkle™ White Disposable Forceps
Disposable forceps are ideal for grasping, lifting, and moving small objects like fine tissue or solid samples.
Brand: Buerkle™ 5378-0040
This item is not returnable.
View return policy
Description
- The fluted handles ensure optimum and secure handling as well as precise work
- The light plastic forceps for single use allow for fatigue-free work
- The clean room-manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined
- The lab tweezers are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses
- Sterile forceps are individually wrapped
- For single use
- Cleanroom manufactured (clean room class 7)
- Complies with EU food requirements and FDA regulations
- Pointed/angled tip: The disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends
- Pointed tip: The anatomical forceps have a straight tip, fluted on the inside. This makes it possible to grip sensitive materials reliably
- Broad tip: The disposable forceps have ends with a broad tip. The broad tip is smooth and blunt. It prevents the material from being damaged when it is gripped
Specifications
Angled, Pointed | |
5.12 in. | |
130 mm | |
Non-sterile |
100 (large pack) | |
Polystyrene | |
Forceps | |
Standard |
Brochure LaboPlast SteriPlast scoops spoons spatulas
File size: 608.8 kB
Topsellers disposamplers
File size: 1.3 MB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
244556 | Certificate of Compliance | 19/11/2024 | 5378-0040 |
241419 | Certificate of Compliance | 11/06/2024 | 5378-0040 |
233475 | Certificate of Compliance | 16/01/2024 | 5378-0040 |
235109 | Certificate of Compliance | 23/11/2023 | 5378-0040 |
230282 | Certificate of Compliance | 20/01/2023 | 5378-0040 |
Product Content Correction
Product Title
Buerkle™ White Disposable Forceps > Non-sterile, Pointed, Angled