Learn More
Whirl-Pak™ Polyethylene Write-on Bags
Brand: Whirl Pak B01490WA
1308.97 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Specifications
384 mL |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Product Content Correction
Product Title
Whirl-Pak™ Polyethylene Write-on Bags > Capacity: 384mL