missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps for Narrow-Mouth Containers
Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
13-425
24-410
28-410
38-430
No. per Case:
48 Cs.
72 Cs.
Unit Size:
Pack of 48
Pack of 72
Description
- Leak-proof design
- Autoclavable
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| No. per Case | 72 Cs. |
| Neck Finish | 13-425 |
| Certifications/Compliance | ISO 9001 |
| For Use With (Equipment) | Redi-pak™ leak resistant narrow mouth bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction