Learn More
DWK Life Sciences Wheaton™ Columbia Jars for Cover Slips
For staining applications, cleaning cover slips, as well as coating them with materials such as poly-lysine or silane. Wheaton™ Columbia Jars for Cover Slips are manufactured from soda lime glass.
Brand: DWK Life Sciences W900180
345.45 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
493.56 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
- Longer slip covers can be accommodated if the cap is removed
- Includes a 43-400 white polypropylene screw cap with a PTFE coated polyethylene liner
- Holds up to 4 cover slips 17-23mm wide and up to 30mm long
Specifications
White | |
Polypropylene | |
Round | |
Staining Jar |
Closure Included | |
Screw | |
Soda Lime Glass | |
4 |
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13