Learn More
DWK Life Sciences Wheaton™ Snap Plug Caps
Manufactured from Polyethylene
Brand: DWK Life Sciences 242806
706.84 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Closure Style: Snap Cap
- Liner Material: TFE
Specifications
Green | |
Polyethylene | |
Plug | |
8 mm |
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Snap Plug Caps > Green; 8mm