Learn More
DWK Life Sciences Wheaton™ Arsine Scrubber Tube With 24/40, 12/2 Joint
Engineered for arsine analysis using the silver diethyldithiocarbamate (SDDC) colorimetric method
Brand: DWK Life Sciences 180022
644.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
902.08 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
- Scrubber tube
- Standard taper 24/40
- Clear-Seal joint and a spherical 12/2 ball joint held together by a size 12 stainless steel ball joint clamp
- Manufactured from WHEATON 33 low extractable borosilicate glass
Compliance
Meets ASTM / USP and EPA specifications; Borosilicate glass conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Specifications
Arsine Scrubber Tube | |
Borosilicate Glass | |
For arsine analysis using SDDC method | |
Size 12 stainless steel ball joint clamp |
24/40, 12/2 | |
USP Type I and ASTM E438 Type I, Class A | |
For arsine generator |
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14