missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ PVC Dropper Tips
Click to view available options
Material:
PVC
Description
- White polypropylene overcap
- Fits all 20mm serum finish containers
Specifications
Specifications
Color | White |
Material | PVC |
No. per Case | 100 Cs. |
Certifications/Compliance | ISO 9001 |
Product Type | Dropper Tip |
For Use With (Equipment) | 20mm Serum finish container |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'