Learn More
DWK Life Sciences Wheaton™ Microlink Caps
Open top caps are ideal for storing and containing volatile solvents, sensitive compounds and corrosive chemicals
Brand:  DWK Life Sciences 240106
 View more versions of this product
Product Code. 11501235
  • 1315.00 NOK / Pack of 50
Only null left
Estimated Shipment: 10-11-2025
Add to basket
Description
Description
Open top caps are ideal for storing and containing volatile solvents, sensitive compounds and corrosive chemicals
  • Polypropylene/silicone/PTFE liner is ultrasonically welded to each cap to help eliminate possibility of toxicity
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black
50 Cs.
ISO 9001
Bottle
Polypropylene
33-430
Autoclavable
Polypropylene/Silicone
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Microlink Caps > Closed top; 33mm-430 Thread; Black