Learn More
DWK Life Sciences Wheaton™ Micro Kit and Accessories
Product Code. 12485432 Shop All Wheaton Science Products Products
Click to view available options
Product Type:
Claisen Adapter
Connecting Adapter
Unit Size:
Each
Product Code. 12485432
Brand: DWK Life Sciences 123753
Only null left
Add to basket
Add to basket
WHEATON standard taper Clear-Seal™ grease-free joints provide smooth, unground surfaces, which seal without grease and reduce seizing and contamination from grease
  • Apparatus for a wide range of organic chemistry experiments at a lower price than purchasing the components separately
  • Clear-Seal™ joints can replace ground units in any application and provide other important advantages over ground joints
  • Superior in vacuum and radioactive applications
  • Smooth surfaces resist contamination
  • Cleaning is simple and thorough
  • Interchangeable with ground joints
  • All chemical changes and thermometer scales remain visible through the joints
  • Clear-Seal joints have never been weakened by grinding and are formed from special wall tubing that is heavier than standard wall for extra strength
  • Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to ASTM E 438 Type I, Class A requirements
  • Clear-Seal joints are produced to American Standard Taper dimensions and can be used interchangeably with any standard taper ground joints
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Wheaton micro kit
Includes 19/22 Clear-Seal™ joints
Product Type Connecting Adapter