Learn More
DWK Life Sciences Wheaton™ Leak-Resistant Amber HDPE Bottles with Caps Product Code.: 10225022
Estimated Shipment: 30-10-2025
Only null left
Add to basket
DWK Life Sciences Wheaton™ Leak-Resistant Amber HDPE Bottles with Caps
Product Code. 10225022 Shop All Wheaton Science Products Products
15 mL, 20-410
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Neck Finish:
13-425
20-410
24-410
28-410
38-430
Product Code. 10225022
Brand: DWK Life Sciences 209124
Only null left
Estimated Shipment: 30-10-2025
Add to basket
Only null left
Add to basket
Ideal primary packaging solution for assays in diagnostic kits, and also chemical and storage purposes. With double-seal closure with cap attached.
With double-seal closure with cap attached
Ideal for short- and long-term storage and packaging.
  • Narrow-mouth
  • No drip pour lip for clean pouring
  • Polypropylene cap attached remains tight over an extended time
  • Convenience packed
TRUSTED_SUSTAINABILITY
Specifications
Capacity 0.5 oz., 15 mL
Capacity (Metric) 15 mL
Capacity (English) 0.5 oz.
Color Amber
Diameter (English) 0.98 in.
Material HDPE
Neck Finish 20-410
Certifications/Compliance ISO 9001
Height (Metric) 56 mm
Height (English) 2.2 in.
Closure Material Polypropylene
Closure Size 20-410 mm
Diameter (Metric) 25 mm
Closure Type Screw Cap
Product Title
DWK Life Sciences Wheaton™ Leak-Resistant Amber HDPE Bottles with Caps >