Learn More
DWK Life Sciences Wheaton™ Kuderna-Danish Evaporative Concentrator With Clear-Seal™ Joints
For concentration of trace amounts of analyte in organic solvent
Brand: DWK Life Sciences 297883
3131.42 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Clear seal joint
Specifications
Kuderna-Danish Evaporative Concentrator | |
Clear seal joint | |
Concentration of trace amounts of analyte in organic solvent | |
ASTM E 438 |