Learn More
DWK Life Sciences Wheaton™ Glass Sample Vials in Lab File without Caps
Packaged in partitioned trays, without caps
Brand: DWK Life Sciences 224814
2087.49 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
Partitioned trays provide an easy way to inventory samples or to store empty vials.
- Autoclavable
- Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
- Amber vials made from borosilicate glass that protects light-sensitive products
Order Info
Caps available separately
Compliance
Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements. Amber vials conform to USP Type 1 requirements for light transmission.
Specifications
Amber | |
Borosilicate Glass | |
Partitioned Tray | |
2.2 dr. | |
Screw Cap | |
USP Type I | |
M-T File™ and Vial Rack |
17 mm | |
8 mL | |
Screw | |
Autoclavable | |
60 mm | |
Sample Vial in Lab File |
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14