Learn More
DWK Life Sciences Wheaton™ Decappers
These crimpers are used for flip cap seals and for aluminum seals
Brand: DWK Life Sciences 224357
This item is not returnable.
View return policy
Description
Specifications
30 mm Caps | |
Hand-operated Decapper | |
Flip Cap Seals and Aluminum Seals |
Product Suggestions
Customers who viewed this item also viewed.
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Decappers > Decapper; Hand-operated; For 30mm caps