Learn More
DWK Life Sciences Wheaton™ Decappers
These crimpers are used for flip cap seals and for aluminum seals
Brand: DWK Life Sciences 224372
1114.91 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
1142.67 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
13 mm Caps | |
Plier-style Decapper | |
Flip Cap Seals and Aluminum Seals |
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Decappers > Decapper; Pliers; For 13mm caps