Learn More
DWK Life Sciences WHEATON™ COMPLETEPAK MODPAK Amber Kit
Range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
85065.00 NOK - 207520.00 NOK
Specifications
Color | Amber |
---|---|
Sterility | Sterile |
Disposable | Single-use |
For Use With (Application) | Compounding pharmaceuticals, Aseptic filling of injectable drug products, Drug discovery, Drug research, Freeze drying / lyophilized drug products, Large and small molecule drug packaging |
Graduated | No |
Product Code | Brand | Diameter (Metric) Outer | Closure Color | Closure Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Closure Color | Closure Type | Quantity | Price | Quantity & Availability | |||
17992661
|
DWK Life Sciences
W991000A-CPMP |
Blue | Ultra Pure Serum Stopper | 278/Pk. |
180845.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
17972661
|
DWK Life Sciences
W991001A-CPMP |
Blue | Omni Flex Serum Stopper | 278/Pk. |
198020.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
17952581
|
DWK Life Sciences
W991002A-CPMP |
Blue | Omni Flex Igloo Lyo Stopper | 278/Pk. |
205095.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
17902671
|
DWK Life Sciences
W991003A-CPMP |
Red | Ultra Pure Serum Stopper | 278/Pk. |
180845.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
17982581
|
DWK Life Sciences
W991004A-CPMP |
Red | Omni Flex Serum Stopper | 278/Pk. |
198020.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
17912671
|
DWK Life Sciences
W991005A-CPMP |
Red | Omni Flex Igloo Lyo Stopper | 278/Pk. |
207520.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
17942671
|
DWK Life Sciences
W991006A-CPMP |
22 mm | Blue | Ultra Pure Serum Stopper |
85065.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
17932671
|
DWK Life Sciences
W991007A-CPMP |
22 mm | Blue | Omni Flex Serum Stopper |
95675.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
17922671
|
DWK Life Sciences
W991008A-CPMP |
22 mm | Blue | Omni Flex Igloo Lyo Stopper |
95185.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
17952591
|
DWK Life Sciences
W991009A-CPMP |
22 mm | Red | Ultra Pure Serum Stopper |
86065.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
17972671
|
DWK Life Sciences
W991010A-CPMP |
22 mm | Red | Omni Flex Serum Stopper |
95675.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
17952601
|
DWK Life Sciences
W991011A-CPMP |
22 mm | Red | Omni Flex Igloo Lyo Stopper |
95185.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||
17962671
|
DWK Life Sciences
W991012A-CPMP |
24 mm | Blue | Ultra Pure Serum Stopper | 104/Pk. |
86120.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
|||
17952671
|
DWK Life Sciences
W991013A-CPMP |
24 mm | Blue | Omni Flex Serum Stopper | 104/Pk. |
109605.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
|||
17982671
|
DWK Life Sciences
W991014A-CPMP |
24 mm | Blue | Omni Flex Igloo Lyo Stopper | 104/Pk. |
112360.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
|||
17992671
|
DWK Life Sciences
W991015A-CPMP |
24 mm | Red | Ultra Pure Serum Stopper | 104/Pk. |
85115.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
|||
17992571
|
DWK Life Sciences
W991016A-CPMP |
24 mm | Red | Omni Flex Serum Stopper | 104/Pk. |
107460.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
|||
17912681
|
DWK Life Sciences
W991017A-CPMP |
24 mm | Red | Omni Flex Igloo Lyo Stopper | 104/Pk. |
110155.00 NOK
1 set |
Estimated Shipment: 04-08-2025
Log in to see stock. |
|||
Description
- Six inventory matched kits conveniently packed into a single shipper case.
- Sterilized, off-the-shelf, ready-to-use (RtU) primary packaging components
- Focuses your effort on core competencies and reduces lead time
- Streamlines process and frees up valuable clean room space USP /EP 2.9.19
- Particulate Matter in Injections, USP /EP 2.6.1,
- The Bacterial Endotoxins Test and USP / EP 2.6.14
- The Sterility Test, USP /EP 3.2.1
- Glass Containers for Pharmaceutical Use
- USP /EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
- Complete manufacturing & certificate of analysis traceability
- Ready-to-Use, Just-in-Time
- Minimum 180 day shelf life from time of shipment
- Container Closure Integrity (CCI) validation
- Available in amber for photosensitive contents
Specifications
Amber | |
Single-use | |
No | |
CompletePAK ModPAK | |
Autoclavable | |
Serum Vial |
Sterile | |
Compounding pharmaceuticals, Aseptic filling of injectable drug products, Drug discovery, Drug research, Freeze drying / lyophilized drug products, Large and small molecule drug packaging | |
Vial, Stopper, and Seal | |
Bromobutyl | |
USP <788>/EP 2.9.19 Particulate Matter in Injections, USP <85>/EP 2.6.1, The Bacterial Endotoxins Test and USP <71>/ EP 2.6.14 The Sterility Test, USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations |
Product Title
DWK Life Sciences WHEATON™ COMPLETEPAK MODPAK Amber Kit