Learn More
DWK Life Sciences Wheaton™ 190mm Soxhlet Extractor With 24/40 Bottom Inner Joint and 34/45 Top Outer Joint
For continuous extraction of analytes from solid into organic solvent
Brand:  DWK Life Sciences 415000
2031.72 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12644823
  • 2885.00 NOK / Each
Estimated Shipment: 13-10-2025
Explore more special offers
Description
  • For Soxhlet extraction apparatus
  • Wetted parts manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
  • Size: 30 x 190mm
  • Case of one
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Soxhlet Extraction Apparatus
Soxhlet Extraction Apparatus
Used for continuous extraction of analytes from a solid into an organic solvent
ASTM E 438
Documents
One moment while we fetch your results.
Product Title
DWK Life Sciences Wheaton™ 190mm Soxhlet Extractor With 24/40 Bottom Inner Joint and 34/45 Top Outer Joint > For Soxhlet extraction apparatus; Size: 30 x 190mm