Learn More
Cytiva Whatman™ GV 100 Series Glass Vacuum Filtration Manifolds, Replacement Parts
Replacement parts for Whatman™ GV 100 Series Vacuum Filtration Manifolds, includes funnels, lower base supports, frits/sieves, centering rings, and clamps.
Brand: Cytiva 10443001
2379.75 NOK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Description: Replacement Part for GV 100 Series Glass Vacuum Filtration Manifolds
- For Use With (Application): Vacuum Filtration
Specifications
Replacement Part for GV 100 Series Glass Vacuum Filtration Manifolds | |
Borosilicate Glass | |
1 Ea. | |
500 mL |
Manifold Model GV 100/0 and GV 100/1 | |
Vacuum Filtration | |
Funnel (500 mL) |