Learn More
Sartorius™ Weighing Platform
Brand: Sartorius™ 6566-50
755.96 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Specifications
Aluminum | |
Weighing Platform | |
52 g |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%