Learn More
Hanna Instruments™ Waterproof pH/Conductivity/TDS Tester
Makes measuring pH quick and easy. Hanna Instruments™ Waterproof pH/Conductivity/TDS Tester is lightweight and waterproof offering high accuracy pH, EC/TDS and temperature measurements in a single handheld unit.
Brand: Hanna Instruments™ HI98129
2578.27 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Combo testers designed for high accuracy measurements using one meter
- Easy to read dual LCD, pH/°C or TDS/°C
- Automatic temperature compensation
- Automatic calibration
- 'Hold' function
- Automatic shut off
- Replaceable pH electrode
Specifications
±2% | |
±2% | |
Yes | |
LCD | |
0 to 14 | |
0.01 | |
HI98129 | |
1ppm (mg/L) | |
±0.5°C | |
32.0°F to 140.0°F |
±0.05 | |
100 hr. | |
Automatic, at one or two points with two sets of standard buffers (pH 4.01 / 7.01 / 10.01 or pH 4.01 | |
0 to 3999 ÎĽS/cm | |
0 to 2000 ppm (mg/L) | |
0.0°C to 60.0°C | |
1ÎĽS/cm | |
0 to 2000ppm (mg/L) | |
pH: automatic; EC/TDS: automatic with β adjustable from 0.0 to 2.4% /°C | |
0.1 / 0.1°C/°F |