Learn More
Water R Solution, Fisher Chemical™ Product Code.: 12940564
Water R Solution, Fisher Chemical™
BIOPHARMA
Product Code. 12940564 Shop All Fisher Chemical Products
5000 mL, 5000mL
Click to view available options
Quantity:
5000 mL
Unit Size:
5000mL
Product Code. 12940564
Brand: Fisher Chemical RP 226 5000
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Quantity 5000 mL
Physical Form Liquid
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number2506015 Certificate TypeCertificate of Analysis Date10/06/2025 Product CodeRP 226 5000
Lot Number2505011 Certificate TypeCertificate of Analysis Date12/05/2025 Product CodeRP 226 5000
Lot Number1108131 Certificate TypeCertificate of Analysis Date28/04/2025 Product CodeRP 226 5000
Lot Number1047582 Certificate TypeCertificate of Analysis Date03/03/2025 Product CodeRP 226 5000
Lot Number1034665 Certificate TypeCertificate of Analysis Date03/03/2025 Product CodeRP 226 5000
Product Content Correction
Product Title
Water R Solution, Fisher Chemical™ >