Learn More
Water Nitrate Free, Chem Lab™ Product Code.: 13472759
Water Nitrate Free, Chem Lab™
Product Code. 13472759
1000 mL, 1 L
Click to view available options
Quantity:
1000 mL
Unit Size:
1 L
Product Code. 13472759
Brand: Chem Lab CL07.2105.1000
Specifications
pH ±7
Color Colorless
Boiling Point 100°C
Odor Odorless
Quantity 1000 mL
Physical Form Liquid
Product Content Correction
Product Title
Water Nitrate Free, Chem Lab™ >