Learn More
Water, for HPLC-MS, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.50 L
Description
Water, for HPLC-MS
Specifications
Specifications
Packaging | Amber Glass Bottle |
Quantity | 1 L |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
2541507 | Certificate of Analysis | 16/06/2025 | W/0112/17,W/0112/15,W/0112/15X,W/0112/17X |
2540912 | Certificate of Analysis | 11/06/2025 | W/0112/17,W/0112/15,W/0112/15X,W/0112/17X |
2539869 | Certificate of Analysis | 09/06/2025 | W/0112/17,W/0112/15,W/0112/15X,W/0112/17X |
2536798 | Certificate of Analysis | 24/05/2025 | W/0112/17,W/0112/15,W/0112/15X,W/0112/17X |
2536175 | Certificate of Analysis | 24/05/2025 | W/0112/17,W/0112/15,W/0112/15X,W/0112/17X |
Safety and Handling
Product Identifier
- Water
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59% -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
Product Content Correction
Product Title
Water, for HPLC-MS, Fisher Chemical™ >