Learn More
Water, HPLC for Gradient Analysis, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Plastic Coated Glass Bottle
Unit Size:
1 L
2.50 L
Specifications
Specifications
Packaging | Amber Glass Bottle |
Quantity | 1 L |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
2541507 | Certificate of Analysis | 16/06/2025 | W/0106/17Z,W/0106/10RSS,W/0106/15X,W/0106/17X,W/0106/PB17,W/0106/17,W/0106/15,W/0106/17A,W/0106/25RSS |
2540912 | Certificate of Analysis | 11/06/2025 | W/0106/17Z,W/0106/10RSS,W/0106/15X,W/0106/17X,W/0106/PB17,W/0106/17,W/0106/15,W/0106/17A,W/0106/25RSS |
2539869 | Certificate of Analysis | 09/06/2025 | W/0106/17Z,W/0106/10RSS,W/0106/15X,W/0106/17X,W/0106/PB17,W/0106/17,W/0106/15,W/0106/17A,W/0106/25RSS |
2540246 | Certificate of Analysis | 09/06/2025 | W/0106/17Z,W/0106/10RSS,W/0106/15X,W/0106/17X,W/0106/PB17,W/0106/17,W/0106/15,W/0106/17A,W/0106/25RSS |
2539391 | Certificate of Analysis | 09/06/2025 | W/0106/17Z,W/0106/10RSS,W/0106/15X,W/0106/17X,W/0106/PB17,W/0106/17,W/0106/15,W/0106/17A,W/0106/25RSS |
Safety and Handling
Product Identifier
- Water
Product Content Correction
Product Title
Water, HPLC for Gradient Analysis, Fisher Chemical™ >