Learn More
Karl Hecht™ Watch Glasses
These watch glasses are useful for evaporating liquids, holding solids, and heating small amounts of a sample.
19.00 NOK - 23.00 NOK
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
11371395
|
Karl Hecht™
40010006 |
6 cm |
19.00 NOK
Each |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
11381395
|
Karl Hecht™
40010010 |
10 cm |
23.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
Description
- Acc. to DIN 12 341
- With fused rim
- Manufactured from glass
- Available in different diameters
Specifications
Fused rim | |
Glass | |
Round |
Product Title
Karl Hecht™ Watch Glasses