Learn More
BRAND™ Watch Glass
Watch Glass constructed of soda-lime glass with ground edges.
402.00 NOK - 600.00 NOK
Specifications
Description | Stress reduced edges |
---|---|
Material | Soda Lime Glass |
Edges | Ground |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
10069980
|
BRAND™
150010 |
40 mm |
402.00 NOK
Pack of 10 |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
10479522
|
BRAND™
150060 |
120 mm |
600.00 NOK
Pack of 10 |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
Description
- Soda-lime glass
- Ground edges
- Stress-reduced to minimize risk of shattering
Specifications
Stress reduced edges | |
Soda Lime Glass | |
Ground |
Product Title
BRAND™ Watch Glass